Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ready-to-use DNA ladders, Biotium
Supplier: Biotium
31032 31022
71003-998EA 126.52 USD InStock
71003-998 89493-044
Ready-to-use DNA ladders, Biotium
Electrophoresis Reagents Molecular Weight Markers
DNA Ladders are generated from PCR followed by restriction enzyme digestion, phenol extraction, and equilibration to 10 mM Tris-HCl (pH 8.0) and 1 mM EDTA
Ready-to-use DNA ladders are supplied in 1X loading buffer at an optimal concentration for loading GelRed™ and GelGreen™ precast gels, and include a vial of 6X loading buffer to add to your DNA samples.
The 1 kb ladder has 13 bands suitable for sizing linear double-stranded DNA fragments from 250 bp to 10 kb. The 1 kb and 3 kb bands have increased intensity to provide internal orientation. The 100 bp DNA Ladder has 11 bands suitable for sizing linear double-stranded DNA fragments from 100 bp to 1500 bp. The 500 bp and 1,500 bp bands have increased intensity to provide internal orientation. The ladders are supplied with a vial of 6X loading buffer, which contains density agents and two blue electrophoresis tracking dyes that run at approximately 1.5 kb and 200 bp in a 1% agarose gel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty