Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Antibody (CF™ 594)

Supplier: Biotium

20118-1MG
89493-220EA 213.98 USD InStock
89493-220
Anti-IgG Goat Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: CF™ 594
Epitope:
Host: Goat
Isotype:
Reactivity: Guinea Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty