Would you like to log in again?
Anti-Avi-Tag Mouse Monoclonal Antibody
Supplier: Genscript
A01738-200 A01738-40
89493-824EA 103.46 USD InStock
89493-824 89493-822
Anti-Avi-Tag Mouse Monoclonal Antibody
Antibodies
Avi Tag Is A Biotin-Acceptor Peptide, Glndifeaqkiewhe. The 15-Residue Peptide Served As A Substrate Mimic For Biotin Ligase (Bira), Which Usually Recognizes The Much Larger Protein Domain. Avi Tag Is A Useful Tool In Analysis Of Avi Fusion Proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty