Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cre Recombinase Mouse Monoclonal Antibody [clone: 1D8H3]

Supplier: GenScript Biotech
GenScript®
A00888-40
89494-016EA 154.65 USD InStock
89494-016
Anti-Cre Recombinase Mouse Monoclonal Antibody [clone: 1D8H3]
Antibodies
Recombinant Cre recombinase from E.coli expression systems.

  • High specificit
  • High affinity
  • High stability

GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.

Type: Tag
Antigen: Cre Recombinase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty