Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Mouse Monoclonal Antibody [clone: 4H3G6]

Supplier: GenScript Biotech
GenScript®
A01730
89494-612EA 94 USD InStock
89494-612
Anti-IgG Mouse Monoclonal Antibody [clone: 4H3G6]
Antibodies
Mouse IgG2b control (Whole Molecule), Purified is suitable to be used as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, immunobloting (Dot blot and Western blot), immunoprecipitation, immunodiffusion, and immunoelectrophoresis. It may also be used as antigen or ligand in immunochemical conjugation reaction. GenScript Mouse IgG2b control (Whole Molecule)is highly purified from normal mouse ascites through Protein A affinity column and produced from the hybridoma resulting from fusion of SP2/0-Ag14 myeloma and B-lymphocytes.

  • High specificit
  • High affinity
  • High stability

GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.

Type: Control
Antigen: IgG2 beta
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty