Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DYKDDDDK Tag Mouse Monoclonal Antibody [clone: 5A8E5] (FITC (Fluorescein Isothiocyanate))

Supplier: GenScript Biotech
GenScript®
A01632
89495-076EA 209.38 USD InStock
89495-076
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody [clone: 5A8E5] (FITC (Fluorescein Isothiocyanate))
Antibodies
GenScript THE™ DYKDDDDK Tag antibody [FITC], mAb, Mouse recognizes C-terminal, N-terminal and internal DYKDDDDK tagged fusion proteins.

  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty