Would you like to log in again?
WHEATON® Headspace Screw Thread Vials and Closures, DWK LifeSciences
Supplier: DWK Life Sciences
W225287 W225284 W225285 W225286
89495-264CS 53.28 USD InStock
89495-264 89495-268 89495-266 89495-270
WHEATON® Headspace Screw Thread Vials and Closures, DWK LifeSciences
Vials
Headspace vials are manufactured from clear Type I borosilicate glass.
  • Clear or amber color
  • Screw thread finish eliminates the need for crimping tools
  • Accepts 18 mm screw thread closures with septa
Ordering information: Caps not included (sold separately).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty