Would you like to log in again?
C-MAG HS 10 IKAMAG® Hot Plate Stirrer, IKA® Works
Supplier: IKA
IKAMAG®
0003581401
89495-320EA 1912.99 USD InStock
89495-320
C-MAG HS 10 IKAMAG® Hot Plate Stirrer, IKA® Works
Stirrers Magnetic Hotplate Stirrers
New magnetic stirrer features heating capabilities and a ceramic heating plate that offers excellent chemical resistance.
Units are equipped with a powerful motor for stirring quantities of up to 15 L (H₂O). Standard safety features include a fixed safety circuit of 550 °C, Hot Top indicator for hot surface warning to prevent burns, a digital error code display, and an elevated control panel for protection against leaking liquids. Exact temperature setting is provided via digital display (LED). Also provided is a bushing according to DIN 12878 for connecting a contact thermometer (see 89028-114 series) to enable precise temperature control.
Units meet IP 21 protection class according to DIN EN 60529.
Certifications: CE marked. UL and C-UL listed.
Ordering information: Supplied with a three-year manufacturer's warranty (2+1 with product registration at www.ika.com/register). For additional laboratory accessory options, see the IKA® Lablift Manual (see 89495-302 series). Accessories are sold separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty