Would you like to log in again?
VWR® Extreme Temperature Tape
Supplier: VWR
VWPT-34AM VWPT-12AM VWPT-1AM
89495-452CS 2925.24 USD InStock
89495-452 89495-450 89495-448
VWR® Extreme Temperature Tape
Tapes
This Extreme Temperature Tape is a transparent amber polyimide film with pressure sensitive silicone adhesive.
  • Transparent amber polyimide film
  • Wound on a plastic core
The extreme temperature-resistant properties (-100°F to 500°F) of this tape make it suitable for high heat and sub zero applications
Recommended applications include flame-resistent electrical insulation, solder mask for circuit boards, heat sealing, lamination, cold storage, lyophilizing, freeze drying, and over-labeling.
The silicone adhesive ensures a clean release from most substrates even after exposure to extreme temperatures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty