Would you like to log in again?
Fluid-Resistant Barrier Coats
Supplier: CritiCore Protective Wear
89495-718 89495-722 89495-726 89495-720 89495-732 89495-728 89495-724 89495-716 89495-730 89495-734
89495-718EA 39.95 USD InStock
89495-718 89495-720 89495-730 89495-724 89495-734 89495-722 89495-732 89495-728 89495-716 89495-726
Fluid-Resistant Barrier Coats
Workwear
These specialized lab coats offer excellent fluid resistance while maintaining their breathability.
  • Fluid-resistant, 100% Polyester front and sleeves
  • PolyCotton back panel for ventilation
  • Quick-release snap front for fast removal
  • Safety-sleeve knit cuff
  • Two hip pockets and one breast pocket
These coats are extremely durable and able to hold up to repeated industrial launderings.
Ordering information: Coats in sizes XS to XXL are stocked. For sizes 3XL to 6XL, extended lead time may be required. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty