Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CritiCore Cleanroom Frocks with ESD Stripe
Supplier: CritiCore Protective Wear
CritiCore
89495-754 89495-756 89495-758 89495-744 89495-766 89495-764 89495-738 89495-748 89495-768 89495-760 89495-746 89495-752 89495-740 89495-762 89495-770 89495-736 89495-750 89495-742
89495-760EA 90.83 USD InStock
89495-760 89495-770 89495-764 89495-742 89495-752 89495-740 89495-762 89495-750 89495-768 89495-746 89495-756 89495-744 89495-766 89495-754 89495-738 89495-748 89495-758 89495-736
CritiCore Cleanroom Frocks with ESD Stripe
Workwear
Reusable zippered frocks are designed for controlled environments and less critical cleanroom applications
  • Garments feature a mandarin collar with two snaps at top, a wide-pull zipper, and single snap below at bottom
  • Assembled with double-needle felled seams for added durability and particle migration prevention
  • All seams are laser-cut or pre-serged, and thread is silicon-free
  • Available in white and navy
Made with lightweight yet durable high-density ESD striped fabric, they resist fluid and bacterial penetration. A 1% carbon stripe provides electrostatic dissipation, and knit cuffs include 5% carbon Belltron fiber. Applications include pharmaceutical, bio-tech, and medical device manufacturing.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty