Would you like to log in again?
CritiCore Cleanroom Coveralls with ESD Stripes
Supplier: CritiCore Protective Wear
CritiCore
89495-800 89495-782 89495-806 89495-786 89495-774 89495-798 89495-794 89495-776 89495-796 89495-792 89495-784 89495-788 89495-804 89495-780 89495-790 89495-772 89495-802 89495-778
89495-806EA 183.7 USD InStock
89495-806 89495-782 89495-792 89495-780 89495-790 89495-786 89495-774 89495-796 89495-784 89495-794 89495-772 89495-800 89495-778 89495-788 89495-798 89495-776 89495-804 89495-802
CritiCore Cleanroom Coveralls with ESD Stripes
Workwear
Reusable cleanroom coveralls are made with a high-density polyester calendered fabric that resists fluids and bacterial penetration.
Coveralls include knit cuffs with 5% carbon Belltron fiber, a full-length, lint-limiting front zipper with two top snaps at neck, and a mandarin collar. The bottom of the legs include three male snaps and one female snap for adjustment, plus three male snaps on the back of the leg for boot attachment. All seams are laser-cut or pre-serged. Thread is silicon-free.
Available in white and navy.
Designed for cleanroom applications, the garments also include a 1% carbon stripe for electrostatic dissipation. Applications include pharmaceutical, bio-tech, and medical device manufacturing.
Certifications: ISO Class 4.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty