Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CritiCore ESD Stripe Cleanroom Hoods with Open Face Below Lip
Supplier: CritiCore Protective Wear
CritiCore
89495-810 89495-808
89495-808EA 46.52 USD InStock
89495-808 89495-810
CritiCore ESD Stripe Cleanroom Hoods with Open Face Below Lip
Workwear
These reusable cleanroom hoods are made with a high-density polyester calendered fabric that resists fluids and bacterial penetration.
  • Hoods are a one-size-fits-all pullover
  • Tunneled elastic back plus horizontal and vertical adjustment snaps
  • Split-drape design with a front panel longer than the back
  • All seams are laser-cut or pre-serged
  • Thread is silicon-free
Designed for cleanroom applications up to and including Class 10 (ISO 4), hoods also include a 1% carbon stripe for electrostatic dissipation. Suitable applications include pharmaceutical, bio-tech, and medical device manufacturing.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty