Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CritiCore Molded Sole Cleanroom Boots with ESD Stripe
Supplier: CritiCore Protective Wear
CritiCore
89495-813 89495-827 89495-815 89495-829 89495-831 89495-823 89495-821 89495-819 89495-817 89495-835 89495-825 89495-833
89495-819PK 82.39 USD InStock
89495-819 89495-829 89495-817 89495-823 89495-833 89495-821 89495-831 89495-827 89495-815 89495-825 89495-813 89495-835
CritiCore Molded Sole Cleanroom Boots with ESD Stripe
Workwear
With a unique anti-slip tread pattern featuring 30% more traction and a high sidewall, these reusable molded-sole cleanroom boots provide superior performance in both wet and dry environments.
  • Gray molded sole
  • 45.7cm (18") fabric uppers with tunneled elastic at ankle and top
  • Three male snaps and one female snap at top for size adjustment
  • One additional reinforced snap on back of boot for coverall leg attachment
  • Reinforced fabric band is sewn around the perimeter of the sidewall for added durability
  • Buckle includes color-coded webbing for easy size identification
  • All seams are laser-cut or pre-serged
  • Thread is silicon-free
  • 99% polyester, 1% carbon material
They are gamma sterilization stable and made with a tightly woven calendered fabric that has excellent particle control properties. Applications include pharmaceutical, bio-tech, and medical device manufacturing.
They are gamma sterilization stable and made with a tightly woven calendered fabric that has excellent particle control properties. Applications include pharmaceutical, bio-tech, and medical device manufacturing.
Ordering information: Cleanroom boots are available in white or navy, with sizes ranging from small to 3X-large. All cleanroom boots ship in a pack of 2. Extended lead time may be required. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty