Would you like to log in again?
CritiCore Cleanroom Coveralls with ESD Grids
Supplier: CritiCore Protective Wear
CritiCore
89495-884 89495-886 89495-876 89495-894 89495-888 89495-882 89495-892 89495-896 89495-904 89495-900 89495-878 89495-898 89495-874 89495-902 89495-906 89495-890 89495-880 89495-872
89495-906EA 199.03 USD InStock
89495-906 89495-892 89495-880 89495-890 89495-896 89495-874 89495-884 89495-894 89495-872 89495-882 89495-900 89495-878 89495-888 89495-876 89495-898 89495-886 89495-904 89495-902
CritiCore Cleanroom Coveralls with ESD Grids
Workwear
Made with durable ESD grid material and advanced garment construction methods, these reusable coveralls stand up to repeated launderings while still maintaining particle control.
Coveralls include knit cuffs with 5% carbon Belltron fiber, a full-length, lint-limiting front zipper with two top snaps at neck, and a mandarin collar. The bottom of the legs include three male snaps and one female snap for adjustment, plus three male snaps on the back of the leg for boot attachment. All seams are laser-cut or pre-serged. Thread is silicon-free.
Available in white and navy.
Designed for controlled environments that need electrostatic dissipation. The fabric features a 1% carbon grid, and is constructed of a dense twill 100% multi-filament yarn that guards against airborne particles, bacteria, and lint. Applications include micro-electronics and semi-conductor manufacturing.
Certifications: ISO Class 5. IEST RP CC03 compliance.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty