Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cleanroom Hood, ESD Grid, CritiCore Protective Wear
Supplier: CritiCore Protective Wear
89495-932 89495-924 89495-930 89495-928 89495-922 89495-910 89495-918 89495-920 89495-926 89495-914 89495-908 89495-912 89495-934 89495-938 89495-916 89495-936
89495-908EA 32.67 USD InStock
89495-908 89495-918 89495-928 89495-938 89495-916 89495-922 89495-932 89495-910 89495-920 89495-930 89495-926 89495-914 89495-936 89495-924 89495-912 89495-934
Cleanroom Hood, ESD Grid, CritiCore Protective Wear
Workwear
These reusable hoods are made with a high-density non-linting material that is rated for ISO Class 3+ cleanrooms requiring electrostatic dissipation (ESD)
Hoods have a tunneled elastic back plus horizontal and vertical adjustment snaps. They feature a split-drape design, with the front panel longer than the back. All seams are laser-cut or pre-serged. Thread is silicon-free.
They include a 1% carbon grid and have excellent durability and longevity. Suitable applications include micro-electronics, semi-conductor manufacturing, and nano research.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty