Would you like to log in again?
Autoclavable Precision Clean® Breather Bags, PPC Flexible Packaging
Supplier: PPC Flexible Packaging
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Clean® Precision Clean®
STCK0042-001 STCK0043-001 STCK0046-001 STCK0045-001 STCK0044-001 STCK0041-001 STCK0047-001
89496-130CS 2195.47 USD InStock
89496-130 89496-132 89496-134 89496-136 89496-138 89496-126 89496-128
Autoclavable Precision Clean® Breather Bags, PPC Flexible Packaging
Bags Sterilization Pouches
A combination of Tyvek™ and clear, ultraclean film structure aids in the prevention of interior particle contamination.
  • Faster cycle times
  • Molecular bonded seals
  • Pouch suitable for sterilizaiton by steam, EtO, gamma radiation or hydrogen peroxide
  • One side Tyvek™ and reverse side Precision Clean® HD 3-mil clear film
It is designed for products to be used in cleanroom environments that also require sterilization.
Certifications: Certified to NASA™ clean standards by Fisher to meet Level 100 KSC-C-123.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty