Would you like to log in again?
MicroPolx® 2750 Polyester/Nylon Microfiber Knitted Wiper, Berkshire
Supplier: Berkshire
MPX275009091
89496-566CS 1428.34 USD InStock
89496-566
MicroPolx® 2750 Polyester/Nylon Microfiber Knitted Wiper, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
MicroPolx® 2750 is a sealed-edge, cleanroom-laundered wiper recommended for ISO class 4 and above.
  • Used in display and semiconductor industries
  • Low levels of fiber and/or particle contamination
  • Abrasion resistance and is chemically compatible with IPA and other solvents
  • Soft hand and edge for sensitive surfaces
Microfiber yarn increases surface area and provides superior wiping efficiency and sorptive performance.
Applications include cleaning precise optical machinery and tools, delicate process in critical cleanrooms, applying and removing cleaning and disinfecting solutions.
It is composed of a polyester and nylon knitted microfiber / microdenier fabric. This combination of properties provides high level of cleanliness, abrasion resistance, and absorptive performance required for ISO Class 4 and above environments, as well as applications requiring critical control of contaminants
Ordering information: 100 sheets per pack, 10 packs per case (double packed).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty