This wireless monitoring solution for laboratory equipment safeguards the integrity of precious samples by continuously monitoring critical parameters and securely logging data to provide unprecedented peace-of-mind. Featuring audit trail traceability, this solution assists with conformance to 21 CFR Part 11, and other strenuous regulatory requirements for labs operating in regulated industries. System features software, modules, repeaters, receivers, and a 915 Mhz frequency (North America).
Continuous, 24/7/365 real-time monitoring of critical parameters, with intelligent data logging using globally recognized radio frequency transmissions; Temperature, Relative humidity, CO2 concentration, Differential pressure, 4-20 mA output, Alarm relay/dry contacts.
Easy-to-read display of monitored set points directly on module including audible/visual siren, telephone, email, text message, and fax.
Single solution that can grow with you to save time and money. Smart-Vue software can run one lab or an entire enterprise, whether across the street, or across the globe. Smart-Vue is easily expandable, allowing new equipment to be added to the existing systems. It's compatible with multiple brands and types of laboratory equipment. Easy to install, use, and maintain. Simple relocation–maintaining net work configurations–without the inconvenience and cost associated with rewiring.
Smart-Vue protects the quality of your samples by continuously monitoring equipment conditions and will remotely notify you if conditions for sample integrity are compromised. In addition, Smart-Vue’s data logging capabilities assist with compliance to with FDA, GxP and other regulatory requirements.
Ordering information: Smart-Vue Remote Wireless Monitoring Systems are comprised of a single software program, signal receivers and modules. Type of equipment to be monitored, proximity to receiver and building configuration are system variables to consider when building the system and selecting appropriate components. Please contact your Avantor sales representative for assistance and to schedule an on site survey if required. The site survey will map out module location, signal strength, receiver location and IT considerations specifically for your lab.