Would you like to log in again?
CryoStar™ NX50 Cryostats
Supplier: Epredia
CryoStar™
957120H 957120K 957260H 957100L 957120L 957250K 957110L 957130H 957230H 957270K 957230K 957230L 957260K 957100H 957270L 957270H 957130K 957100K 957260L 957110K 957110H 957250H
89497-208EA 50584.6 USD InStock
89497-208 89497-206 89497-204 89497-214 89497-202 89497-212 89497-200 89497-178 89497-188 89497-210 89497-176 89497-198 89497-186 89497-196 89497-174 89497-184 89497-172 89497-194 89497-192 89497-170 89497-180 89497-190
CryoStar™ NX50 Cryostats
Microtomes and Microtome Assemblies Microtomes
The CryoStar™ NX50 Cryostat extends the benefits of the NX series by offering an innovative new look at cryosectioning. The NX series has been designed completely around the user to deliver optimal sectioning quality, unmatched ergonomics, and safety in an energy efficient package. With the NX series, users can deliver high quality results comfortably, safely, and efficiently.
The CryoStar™ NX50 Cryostat follows in the footsteps of the popular CryoStar™ NX70 Cryostat by offering standard features well-suited to the histology laboratory, including Rapid Response Object Head Temperature Control, form-fitting design, and a color touch screen user interface for control of instrument functions
The cryochamber features a spacious stainless steel chamber for workflow organization, chamber cooling to –25 °C, ±2 °C (at 20 °C ambient), separate specimen head cooling to –43 °C, ±2 °C (at 20 °C ambient), and a continuously cooled knife holder to –27 °C, ±2 °C (at 20 °C ambient). Eighteen cryobar storage positions include one quick freeze location. The integrated peltier fast freezing device rapidly cools to –57 °C, ±3 °C. The microtome features a manual light touch rotary flywheel, with sample orientation adjustable 8° in the Y axis for increased stability and 360° Z axis rotation.
Environmental Note: The NX50 and NX70 Cryostats utilize a rapid response cooling system that actively cools all areas of the cryochamber that come in contact with the sample, including the specimen clamp, cryobar, and blade. The result is precise temperature control and half the energy consumption of other routine cryostats. Low energy LED lighting with user-adjustable intensity is also included.
The unit also includes manual sectioning and motorized coarse advance to a maximum of 3 mm/s. Optional features include Integrated Height Control, Vacutome, and Cold D™ Cold Disinfection which disinfects the cold cryochamber in just 50 minutes.
Certifications: Certificate of Hygiene. CE marked.
Ordering information: Standard units include four 30 mm specimen chucks (red, green, blue, and gold), two allen keys (5 and 6 mm), waste tray, brush shelf, three brushes (coarse cleaning brush, magnetic small beveled brush, and magnetic goat's hair brush), blade handling tool, power cord, USB stick, adjusting pin, operator guide, and anti-roll plate. Units with Vacutome include an additional brush for cleaning vacuum hose, coarse filter for use with vacutome, hose extension with nozzle for vacutome, nozzle for sectioning, and filter bags (25 pcs.). Units with Cold Disinfection include an additional measuring cup and Plastic funnel. Note: Low or High Profile Blade holders, or 16cm Steel Knife Carriers are required accessories, and must be designated at the time of ordering.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty