Would you like to log in again?
Superdex 200 Increase Small-Scale SEC Columns, Cytiva
Supplier: Cytiva
Superdex™
28-9909-45 28990946 28990944 28-9909-46 28990945 28-9909-44
89497-276EA 3924.97 USD InStock
CA11029-392 CA11029-394 89497-276 89497-274 89497-272 CA11029-390
Superdex 200 Increase Small-Scale SEC Columns, Cytiva
Chromatography Columns
Superdex 200 Increase size exclusion chromatography columns are well suited for high-resolution analysis and small-scale purification of monoclonal antibodies (mAb) and other biomolecules with Mr~ 10000 to 600000.
  • Enhanced performance: Improved resolution and runtime compared to predecessor Superdex 200
  • Very high resolution: Small bead size and narrow particle size distribution provide high resolution, for high protein purity
  • High flow rates: Rigid beads give excellent pressure/flow properties
Superdex 200 Increase is a next-generation, dextran-agarose composite matrix for SEC. With smaller and more rigid beads than their predecessors, Superdex Increase columns deliver higher resolution purification in shorter run times.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty