Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10% fixative reagent, neutral buffered (10% NBF)

Supplier: AZER SCIENTIFIC INC
Danger
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

NBF55G
89497-305EA 748.78 USD InStock
89497-305
Formalin 10% fixative reagent, neutral buffered (10% NBF)
Formalin
Azer Scientific's 10% neutral buffered formalin fixative is ideal for use as
a general purpose fixative for all tissues.

  • General Purpose Fixative for All Tissues
  • Light Microscopy and Immunhistochemistry Optimizes Histologic Results
  • Ten Percent Neutral Buffered Formalin

Our formalin penetrates quickly but fixes slowly.

It is enhanced by buffering that optimizes histologic results by light microscopy and immunhistochemistry.


Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty