Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
RW 28 Digital Mechanical Overhead Stirrer, 230 V
Supplier: IKA
5040000
89497-436EA 9075.52 USD InStock
89497-436
RW 28 Digital Mechanical Overhead Stirrer, 230 V
Stirrers Overhead Stirrers
This powerful, mechanically controlled laboratory stirrer is designed for highly viscous applications with quantities up to 80L (H₂O). It is suitable for intensive mixing in laboratories and pilot plants. The stirrer comes equipped with overheating protection in the motor by means of self-locking temperature limiter. Additionally, stirring shaft protection and clamping fixture options are available to further secure bowls.
  • 230 V model
Standard features include, digital speed display, infinitely adjustable speed in two speed ranges, push-through agitator shafts, overload protection, and error code display
Units meet IP 40 protection standards according to DIN EN 60529.
Robust, ergonomic design. Quiet operation, with constant power-drive.
Certifications: CE marked.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration at www.ika.com/register). Unit includes chuck, chuck key, and power cord. Stirring elements, boss head clamps, stands, and other accessories are sold separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89497-434) is also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty