Would you like to log in again?
MacConkey Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
K194 K194 K196 K196
89497-612PK 92.69 USD InStock
89497-612 89426-250 CA89426-250 CA89497-612
MacConkey Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
MacConkey Broth available with or without Durham Tube, optically clear, shatter resistant, polycarbonate tube, order by the package of 20, by Hardy Diagnostics.
For the detection of gram-negative, lactose fermenting bacilli. Availabe with a durham tube for gas detection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty