Would you like to log in again?
Letheen Broth, Modified, Hardy Diagnostics
Supplier: Hardy Diagnostics
K293 K293
89497-620PK 109.81 USD InStock
89497-620 CA89497-620
Letheen Broth, Modified, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Letheen Broth, Modified, 16x125mm tube, 9ml fill, order by the package of 20, by Hardy Diagnostics.
Used to determine the antimicrobial activity of quaternary ammonium compounds and in the microbial evaluation of cosmetics.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty