Would you like to log in again?
Extra Long Transfer Pipettes, Globe Scientific
Supplier: Globe Scientific
139030 139050-S20 139038 139050B 139030-S20
76285-418CS 71.97 USD InStock
76285-418 89497-710 76285-406 89497-708 89497-716
Extra Long Transfer Pipettes, Globe Scientific
Pipettes Pasteur/Transfer Pipettes
Produced from a custom grade low-density polyethylene (LDPE), these unbreakable one-piece pipettes are used to draw and dispense liquids in controlled, reproducible drop sizes.
  • Non-toxic and chemically inert to biological fluids and most acids
  • Easy to grip with gloves
  • Temperature Range: –50 to 90 °C
  • Can be gas (EtO) sterilized
Available in 9 and 12" lengths, extra long transfer pipettes are ideal for reaching into large bottles, 24-hour urine containers, jars, and cylinders.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty