Would you like to log in again?
Self Standing Sample Tubes, 5 and 10 ml, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6101B 6101R 6101 6101C 6101G 6101Y 6101S 6102Y 6102C 6102 6102B 6102G 6102S 6102R 6102AM
89497-748CS 238.66 USD InStock
89497-748 89497-736 89497-746 89497-734 89497-756 89497-744 89497-732 89497-754 89497-742 89497-730 89497-752 89497-740 89497-750 89497-728 89497-738
Self Standing Sample Tubes, 5 and 10 ml, Globe Scientific
Tubes Test Tubes
Transport tubes are designed for the safe storage and transport of biological materials.
  • Tubes are manufactured from polypropylene and feature a skirted conical bottom base and molded graduations
  • Caps are manufactured from polyethylene
These transparent polypropylene vials allow unobstructed viewing of the sample. They are shatter-proof and may be exposed to freezing temperatures.
The leak-resistant screw caps can be removed or sealed in a single turn.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty