Would you like to log in again?
Polystyrene Test Tubes, 16×150 mm, Globe Scientific
Supplier: Globe Scientific
110110
89497-776CS 352.39 USD InStock
89497-776
Polystyrene Test Tubes, 16×150 mm, Globe Scientific
Tubes Test Tubes
These tubes are molded from premium-grade polystyrene for clarity.
They offer a spin rate of 1500 rpm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty