Would you like to log in again?
Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
110443 110446 110444 110438 110406 110414 110440 110416 110441 110442 110405 110412 110410LB 110441AM 110410 110409 110428 110410I 110413
89497-814PK 71.49 USD InStock
89497-814 89497-802 89497-812 89497-822 89497-800 89497-810 89497-820 89497-798 89497-796 89497-794 89209-796 10014-244 10014-224 10014-252 10014-242 89497-818 89497-806 89497-816 89497-804
Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Tubes Test Tubes
These tubes are molded from either premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents.
Ordering information: Unless otherwise specified, test tubes are supplied without caps.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty