Would you like to log in again?
Snap Racks for 25 mm Tubes, Globe Scientific
Supplier: Globe Scientific
456704 456703 456706 456710
89498-038EA 38.05 USD InStock
89498-038 89498-044 89498-040 89498-042
Snap Racks for 25 mm Tubes, Globe Scientific
Racks Tube Racks
These innovative polypropylene test tube racks are shipped flat in order to reduce shelf space.
  • Polypropylene (PP)
  • Autoclavable
  • Non-floating in water baths
  • Stackable when empty
  • Ships flat for ease of storage
Snap racks feature molded alphanumeric markings to enable quick and easy test tube identification. Available in assorted colors. Snap racks are non-floating in water baths.
They simply snap together for use. The special design allows all samples to be incubated at the same temperature. The racks can be used in freezers without deforming or becoming brittle. They will not rot, rust, or deform.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty