Would you like to log in again?
Grip Racks with Tube Ejector, Globe Scientific
Supplier: Globe Scientific
456926 456925
89498-060EA 52.34 USD InStock
89498-060 89498-058
Grip Racks with Tube Ejector, Globe Scientific
Racks Tube Racks
These specially designed test tube racks feature rubber grips that hold test tubes in place when decanting liquid.
  • Produced from polyoxymethylene (POM)
  • Holds various size tubes up to 15 mm diameter
  • 50-place rack (5×10 configuration)
  • Dimensions: 260×140×82 mm (10×5.5×3")
  • Autoclavable
The racks also feature a unique tube ejector that allows the user to easily eject all tubes into a waste receptacle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty