Would you like to log in again?
Samco™ Bio-Tite™ Specimen Container, 40 ml/48 mm (1 oz.), Thermo Scientific
Supplier: Thermo Scientific
BioTite™
04 0506 04 0181
89498-390CS 385.4 USD InStock
89498-390 89498-396
Samco™ Bio-Tite™ Specimen Container, 40 ml/48 mm (1 oz.), Thermo Scientific
Sample Containers General Purpose Containers
Prevent bio-hazardous spills with the leak-resistant Samco™ Bio-Tite™ Wide-Mouth 40mL (1oz.) 48mm Specimen Containers
  • Plug seal cap ensures excellent leak-resiatant performance
  • Cap and base knurls ensure ease of handling
  • Supplied nonsterile
These stackable, shatter-resistant containers are manufactured from virgin polypropylene and are ideal for the collection and transportation of both liquid and solid samples. Supplied bulk packed, containers are ideal for pre-fill applications.
Ordering information: Ambient storage and shipping recommended for all containers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty