Would you like to log in again?
BBL™ Tellurite Solution 1%, BD Biosciences
Supplier: BD Biosciences
215294 215294
89498-684PK 789.11 USD InStock
CA11012-396 89498-684
BBL™ Tellurite Solution 1%, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Tellurite solution is a sterile 1% solution of potassium tellurite, a differential agent.
  • Sterile
  • Recommended for the selective isolation of Staphylococci and Corynebacteria
  • 5 mL per vial
Coagulase-positive staphylococci reduce tellurite and produce black colonies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty