Would you like to log in again?
Buffer No. 3, Hardy Diagnostics
Supplier: Hardy Diagnostics
U303 U303
89498-952PK 471.38 USD InStock
CA89498-952 89498-952
Buffer No. 3, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Buffer No. 3, USP<81>, 500ml fill, 500ml polypropylene bottle, order by the package of 10, by Hardy Diagnostics
Buffers to be used in USP <81> antibiotic assay testing.
Buffer No
3, USP<81>, 500ml fill, 500ml polypropylene bottle, order by the package of 10, by Hardy Diagnostics
Certifications: USP<81>
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty