Would you like to log in again?
durAstatic® Dissipative HDPE Bottle, with Take-Along Pump, Menda
Supplier: Menda
35287
89499-032EA 39.19 USD InStock
89499-032
durAstatic® Dissipative HDPE Bottle, with Take-Along Pump, Menda
Bottles Plastic Bottles
Dissipative HDPE durAstatic® bottle protects valuable static sensitive printed circuitry.
  • No migratory additives
  • Embossed with ESD protective symbol
  • Vessel of 6 oz. includes measurements on one side to monitor fluid level
  • Dispenses approximately 0.2 cc of liquid with each pump
  • One-handed operation facilitates dispensing small amount of liquid into top dish
HDPE durAstatic® bottles are dissipative and are designed to protect valuable static sensitive printed circuitry. No migratory additives are used, reducing the chance for contamination from the bottle.
On each take-along Pump there is an opening in the center of the dish to allow fluid to seep back down into the bottle. It is equipped with a locking mechanism. Pump will not leak when lock is engaged, making it suitable for field service kits/travel. Genuine Menda 'dish-type' liquid dispenser pumps and bottle. Stainless steel pump includes spring-loaded lid which will stay open or stay closed to minimize evaporation and odors.
Certifications: Long lasting dissipative properties: <2 second charge decay, tested per IEC 61340-5-1:1998.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty