The dissipative HDPE durAstatic™ bottle is designed to protect valuable static sensitive printed circuitry.
- Available in four colors: orange, green, blue, and yellow
- No migratory additives
- Embossed with ESD protective symbol
- Colored bottles make it easy to identify various chemicals on the workstation
- Printed with bottle contents to eliminate the need for additional labels
No migratory additives are used, to reduce the chance for contamination from the bottle. Colored bottles allow for quick and easy identification in the workstation.
Certifications: Long lasting dissipative properties: Rtt: 1×10⁴ to <1×10¹¹ ohms tested per ANSI/ESD STM11.12; <2 second charge decay, tested per IEC 61340-5-1:1998.