Would you like to log in again?
durAstatic® Dissipative HDPE Bottles, with One-Touch Pump, Menda
Supplier: Menda
35595 35597 35735 35760 35594 35596 35730
89499-068EA 23.83 USD InStock
89499-068 89499-070 89499-074 10147-616 10147-626 89499-072 10147-606
durAstatic® Dissipative HDPE Bottles, with One-Touch Pump, Menda
Bottles Plastic Bottles
The 6 oz. dissipative HDPE durAstatic® bottle with one-touch pump protects static sensitive printed circuitry.
  • No migratory additives
  • Embossed with ESD protective symbol
  • Colored bottles make it easy to identify various chemicals on the workstation
  • Dispenses approximately 0.2 cc of liquid with each pump
  • One-handed operation facilitates dispensing small amount of liquid into top dish
  • Printed with the bottle contents to eliminate the need for additional labels
HDPE durAstatic® bottles are dissipative and are designed to protect valuable static sensitive printed circuitry. No migratory additives are used, reducing the chance for contamination from the bottle.
The one-touch pump is equipped with an opening in the center of the dish to allow fluid to seep back down into the bottle. Genuine Menda 'dish-type' liquid dispenser pumps and bottle. Stainless steel pump includes spring-loaded lid which will stay open or stay closed to minimize evaporation and odors. Printed with the bottle contents to eliminate the need for additional labels. Genuine Made in the United States of America Menda pump and bottle.
Certifications: Long lasting dissipative properties: Rtt: 1×10⁴ to <1×10¹¹ ohms tested per ANSI/ESD STM11.12; <2 second charge decay, tested per IEC 61340-5-1:1998.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty