Would you like to log in again?
Micrewtube™ Microcentrifuge Tubes, Simport Scientific
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
T361-7T T341-6T T361-5TPR T361-2TPR T341-4T T341-6T T361-4T T341-7TPR T341-7T T341-6TPR T361-5T T361-4TPR T361-6TPR T361-2T T341-7TBR T341-4TPR T341-6TBR T341-7TLST T361-5TPR T341-5T T341-4T T341-7TBR T341-7TPR T361-7TPR T341-2TBR T341-2T T341-7T T361-4T T341-5TBR T361-2TPR T361-6T T341-5TBR T361-7T T341-5TLST T361-5T T341-2TPR T341-2TPR T341-4TBR T341-6TLST T341-2TBR T341-7TLST T341-6TBR T341-4TBR T341-5TLST T341-5TPR T341-5T T341-6T T361-4TPR T361-6T T341-2TLST T341-6TLST T341-2TLST T341-6TPR T341-4TLST T341-2T T341-5TPR T361-7TPR T361-6TPR T341-4TLST T341-4TPR T361-2T
89499-104PK 179.6 USD InStock
89499-104 CA11028-582 89499-102 CA11028-580 89499-108 CA11028-542 89499-106 CA11028-540 CA11028-584 CA11012-552 89499-100 60872-412 CA11028-578 CA11028-538 89499-114 89499-112 CA11028-554 89499-118 CA11028-552 89499-116 CA11028-590 89499-078 89499-110 89499-076 CA60872-413 CA60872-412 CA11028-544 CA11028-588 CA11028-548 89499-126 CA11028-560 89499-124 CA73960-270 89499-128 CA11028-562 CA73960-274 89499-084 89499-082 89499-122 89499-088 89499-120 89499-086 CA60872-801 89499-080 CA11028-558 CA11028-556 89050-130 CA60872-802 CA11028-572 CA11028-576 89050-128 CA11028-574 CA11005-666 89499-096 89499-094 89499-098 89499-130 89499-092 89499-090 CA11028-568 CA11028-566
Micrewtube™ Microcentrifuge Tubes, Simport Scientific
Tubes Microcentrifuge Tubes
Micrewtube™ Microcentrifuge tubes are ideal for freezer storage, boiling application and centrifugation, and will fit most standard microcentrifuge rotors.
  • Made of polypropylene
  • Can be used at temperatures from –196 to 121 °C
Low adhesion Micrewtubes™ Tubes are also available. The specially formulated polypropylene used to manufacture these tubes provides a low adhesion surface to obtain maximum sample yield.
Non-skirted tubes withstand high speed centrifugation up to 20000 ×g. Skirted tubes can withstand up to 17000 ×g. Available plain, graduated, or brown for light sensitive material. Available with or without molded ridges.
Certifications: Certified RNase-, DNase-, DNA- and Pyrogen-free.
Ordering information: Caps sold separately; please see T340 Series for caps.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty