Would you like to log in again?
Microcentrifuge Tubes, Graduated, with ClickLok™ MCT Seals, Simport Scientific
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
T330-7B T330-7N T330-8G T330-72G T330-7N T330-8Y T330-72N T330-72AG T330-15G T330-15Y T330-8B T330-8B T330-7AM T330-72N T330-6G T330-8N T330-72Y T330-6G T330-15Y T330-15B T330-72AY T330-72B T330-72AN T330-72AG T330-72G T330-7Y T330-15B T330-72AY T330-7Y T330-6Y T330-7AM T330-6B T330-7B T330-15G T330-72Y T330-72AB T330-15N T330-6N T330-15N T330-7G T330-6B T330-8Y T330-6Y T330-8N T330-72B T330-7G T330-72AM T330-8G T330-72AAM T330-72AM T330-6N T330-72AN
89499-304CS 730.04 USD InStock
89499-304 89499-306 89499-300 89499-302 CA11031-966 CA11031-922 CA11031-924 CA11031-968 CA11031-962 CA11031-964 CA11031-960 89499-278 89499-310 89499-276 CA11031-948 89499-274 CA11031-956 60872-602 60872-604 CA11031-952 60872-601 CA11031-954 CA11031-950 CA60829-035 60872-606 89499-308 89499-326 89499-328 89499-288 89499-322 89499-324 89499-282 89499-280 CA11031-938 89499-286 89499-284 CA11031-946 CA11031-940 CA73960-308 CA73960-306 89499-294 89499-292 89499-298 CA11031-926 89499-296 CA11031-928 CA11031-934 89499-290 CA11031-932 CA11014-052 CA11031-970 CA11031-972
Microcentrifuge Tubes, Graduated, with ClickLok™ MCT Seals, Simport Scientific
Tubes Microcentrifuge Tubes
These polypropylene microcentrifuge tubes are able to withstand the stress of high speed centrifugation up to 20000×g.
  • With external graduation and frosted writing surfaces
  • Autoclavable to 121 °C
  • Pierceable lid
60872-600 incorporates convenient pick-up tabs for easier handling without actually touching the tube. 89499-322 offers a special locking system which can be bent upward to lock on the cap, ensuring extra protection during critical steps.
The one-piece construction incorporates a snug fitting and has a reliable attached cap, even with prolonged boiling. The bottom is reinforced for added protection against leakage.
Certifications: Certified RNase-, DNase-, Pyrogen- and DNA-free.
Ordering information: Packaged in tamperproof, resealable safety-lock bags.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty