Would you like to log in again?
VWR® Flexible Safety Caps
Supplier: VWR
Hexa-Flex™
8380 8360 8365 8370 8375 8350 8355
89499-996PK 62.78 USD InStock
89499-996 89500-000 89499-998 89499-992 89499-994 89499-990 89500-002
VWR® Flexible Safety Caps
Closures Test/Sample Tube Closures
Safety caps provide a safe and economical way to cap and recap blood collection tubes and disposable glass culture tubes, regardless of manufacturer.
  • Caps are color-coded for easy identification
  • Liquid-tight seals
  • Safe and economical
VWR® Flexible Safety Caps are available for a wide range of biomedical and industrial applications and fit six different sized tubes. VWR® Flexible Safety Caps come in a variety of colors for coding samples. They feature liquid-tight seals to ensure sample integrity and prevent spillage of serum samples. Made in the USA.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty