Would you like to log in again?
Autosampler Vial Screw Thread Caps, 9 mm, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
C5000-53A C5000-53R C5000-54G C5000-54Y C5000-56AL C5000-98B C5000-54R C5000-55G C5000-55Y C5000-54A C5000-46M C5000-55P C5000-57B C5000-53Y C5000-53P C5000-57G C5000-55A C5000-55R C5000-53G C5000-47M C5000-54P
89500-044PK 73.61 USD InStock
89500-044 89500-022 89500-032 10033-046 89500-020 89500-042 89500-030 10033-044 89500-040 10033-042 89500-018 89500-028 89500-038 89500-016 89500-026 89500-048 89500-014 89500-036 89500-024 89500-046 89500-034
Autosampler Vial Screw Thread Caps, 9 mm, Thermo Scientific
Closures Chromatography Vial Closures
With Advanced Vial Closure System (AVCS) technology built into these screw caps you will get an optimal, consistent seal every time.
  • Prevent septa push through and over- and under-tightening
With just one turn, these caps offer easy-on, easy-off convenience, and are a cost effective alternative to caps with bonded septa.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty