Would you like to log in again?
Securtainer™ Specimen Containers, Polypropylene, Simport Scientific
Supplier: Simport
C575-60MA C575-20MA C576-60MA C575-90MA C575-20MA C575-40MA C577-20W C577-120W C575-40MA C575-60MA C576-120MA C577-90W C576-20MA C576-40MA C577-60W C577-40W C575-120MA C575-90MA
75857-052CS 160.94 USD InStock
75857-052 89213-164 89213-160 89213-156 CA11027-886 89213-158 CA11027-888 89500-340 75840-150 75840-152 CA11027-884 CA11027-890 CA11027-892 89500-338 89500-336 75840-154 89500-342 75840-156
Securtainer™ Specimen Containers, Polypropylene, Simport Scientific
Sample Containers General Purpose Containers
Securtainer™ Specimen Containers are designed for collection, transport and storage of specimens
Available in three unique models, containers and caps are manufactured without the use of plasticisers or mold release agents. Material used in manufacturing is free from latex. All containers are 95kpa compliant. Available in many sizes from 20 to 120mL. Non-sterile.
The SecurTainer™ I sample container incorporates a unique push tab tamper evident locking mechanism. Once fully screwed, the tamper evident ring will detach itself from the cap showing the container was opened. The Securtainer™ I can also be used without the tamper evident tab.
The SecurTainer™ II sample container incorporates a "soda bottle" type tamper evident mechanism. By simply screwing the cap on the container, the tamper evident sealing ring will automatically lock itself over the container threads. When unscrewed, the ring is detached from the cap showing clearly that the container was opened. 
The SecurTainer™ III sample container is supplied with a standard leakproof screw cap.
Simport Scientific offers shatter-resistant polypropylene containers, eliminating most problems of leakage and evaporation. Containers are uniquely stackable, shatter-resistant and are manufactured from virgin, high-clarity polypropylene. The magenta lids are ribbed for easy opening when hands are wet or gloved while the jars are stackable for easy, safe storage. Containers are translucent to allow specimens to be viewed without opening. Caps are manufactured from virgin polyethylene with an integrated leak-resistant seal. 
Certifications: CE Certified
Ordering information: Containers and caps packaged separately in bags of 100. Color coding CapinserstTM sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty