Would you like to log in again?
Color Frosted Ground Edge Microscope Slide, Globe Scientific
Supplier: Globe Scientific
1324D 1324B 1324P 1324W 1324Y 1324L 1324N 1324T
89500-494CS 885.89 USD InStock
89500-494 89500-482 89500-492 89500-480 89500-490 89500-488 89500-476 89500-486
Color Frosted Ground Edge Microscope Slide, Globe Scientific
Slides Microscope Slides
Color frosted microscope slides are offered in 10 bright, attractive colors.
  • Dimensions: 25×75×1 mm
  • Thickness: 1.1 mm
  • Color frosted one end, one side
  • Packaging: 72/box, 20 boxes/case (10 gross)
Slides are pre-cleaned and supplied ready for use.
All slides are printed with 'Globe' under the frosting area for orientation purposes. These slides feature safety corners, which provide slightly clipped corners for comfortable handling and to prevent the risk of finger and hand cuts.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty