Would you like to log in again?
DiamondⓇ White Glass Microscope Slides, Color Frosted, Globe Scientific
Supplier: Globe Scientific
Diamond™
1380-50D 1380-50A 1380-50W 1380-50Y 1380-50L 1380-50B 1380-50T 1380-50G 1380-50N
89500-528CS 885.06 USD InStock
89500-528 89500-538 89500-526 89500-536 89500-524 89500-534 89500-522 89500-520 89500-530
DiamondⓇ White Glass Microscope Slides, Color Frosted, Globe Scientific
Slides Microscope Slides
DiamondⓇ White Glass microscope slides are made from chemically pure white glass sheets.
  • Produced from the finest quality optical white glass
  • Color coded on one end, one side
  • Packed in high-quality white plastic boxes
  • Dimensions: 25×75 mm
  • Thickness: 1 mm
DiamondⓇ White Glass color coded microscope slides are offered in bright, attractive color coding on one end of each slide. They feature ground edges with 90° corners. The slides are supplied pre-cleaned and ready to use.
The glass offers a low iron content, which results in superior transparency, very low self-florescence, and no color distortion. The special composition of the glass provides minimal oxidation, which ensures sample integrity during long-term storage. Increased hardness and molecular stability of the slide surface guarantees more accurate experimental results.
Ordering information: Slides are packaged 72 per box, 20 boxes per case (10 gross). Minimum order of 10 cases of 1440 for 89500-520, 89500-524 and 89500-534.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty