Would you like to log in again?
Cobalt® Powder-Free Nitrile Examination Gloves, Microflex®
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
N192 N191 N193 N193 N194 N194 N190 N190 N191 N192
75841-204CS 162.76 USD InStock
CA75841-204 75841-204 89500-618 CA89500-620 CA89500-622 89500-624 CA89500-624 89500-622 89500-620 CA89500-618
Cobalt® Powder-Free Nitrile Examination Gloves, Microflex®
Gloves
This soft and stretchy nitrile glove is the best-selling glove from the Microflex® brand of products.
  • Ambidextrous
  • Nonsterile
  • Beaded cuffs
  • Exceptional tactile sensitivity
  • Powder-free
  • Durable and resistant to punctures
Durable and puncture resistant with a fully textured surface for enhanced grip. Ideal for jobs requiring exceptional tactile sensitivity. Gloves are 'cobalt' blue, nonsterile, and ambidextrous. They are also fully textured and feature beaded cuffs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty