Would you like to log in again?
2100Qis with USB+Power Module, Hach
Supplier: Hach
2100QIS01USB 2100QIS01USB
89500-682EA 4208.56 USD InStock
89500-682 CA89500-682
2100Qis with USB+Power Module, Hach
Turbidity Meters
The Hach 2100Qis Portable Turbidimeter offers unsurpassed ease of use and accuracy in turbidity measurement.
  • Portable
  • Easy to use
  • Advanced features
Only Hach offers this unique combination of advanced features and measurement innovation, giving you accurate results every time
The Hach 2100Qis Portable Turbidimeter ensures easy calibration and verification with simple data transfer. It is accurate for rapidly settling samples and provides convenient data logging. Optical System provides precision in the field. USB User interfaces are available in English, French, German, Italian, Spanish, Portuguese (BR), Portuguese (PT), Bulgarian, Chinese, Czech, Danish, Dutch, Finnish, Greek, Hungarian, Japanese, Korean, Polish, Romanian, Russian, Slovenian, Swedish, and Turkish.
Measurement method for 2100Qis is ratio turbidimetric determination using a primary nephelometric light scatter signal (90°) and transmitted light scatter signal. There are three measurement modes including Normal (Push to Read), Signal Averaging, and Rapidly Settling Turbidity. Response time in Normal mode is six seconds. the 2100Qis has an enclosure rating of IP67.
The USB+Power Module provides additional benefit by allowing the user to transfer data to a PC in XML format, transfer data to the Citizen PD-24 printer, operate the instrument on line power (100-240Vac, 50/60 HZ), charge NiMH batteries,perform firmware updates, and quickly enter Sample and User IDs with the optional barcode scanner.
Certifications: CE/RoHS/WEEE, ISO 7027.
Ordering information: Purchase of 2100Qis includes instrument kit and USB+Power Module, case assembly, 4 AA alkaline batteries, 6 sample cells, StablCal ampule kit, silicone oil, oiling cloth, manual. Requires 4, AA Batteries.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty