Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PolyMitt™ Cleaning Mitt Wipes, Micronova

Supplier: Micronova

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

MittWipes™
MWDT-1111IR MW-6 MW-611 MWD6-1211T MW-AA MWDT-AW11 MWDT-1111 MWD-611IR MW11 MW-6IR MWDAA-1211T
89402-994CS 311.94 USD InStock
89402-994 52428-596 89092-746 68400-518 77394-792 68400-678 10029-770 89187-762 89108-090 89501-300 89187-764
PolyMitt™ Cleaning Mitt Wipes, Micronova
Wipes Reusable Cleaning Cloths Cloths for General Applications
These cleaning mitts offer the versatility of a high quality wipe with the convenience and security of a glove. Designed for simplified cleaning around hard to reach areas in the cleanroom.

  • Durable cleaning mitts for simplified cleaning
  • Versatility of a high quality wipe with the convenience and security of a glove
  • Dimensions: 30×20 cm (12×8")
  • For aseptic applications: Available irradiated or can be autoclaved prior to use
  • Available with or without a urethane foam layer
  • Compatible with strong disinfectants
  • Reusable and autoclavable

The Micronova PolyMitt™ Cleaning Mitt Wipes come in one standard size, designed to fit comfortably over a gloved hand, making it easier to clean around carts, shelving, tanks, tubing and other hard to clean areas in the cleanroom.

Available in a range of fabrics for specialized cleaning:
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - 100% textured polyester for residue removal and disinfectant dispersal
MegaTe×™ - 100% non-woven polyamide is absorbent with mild abrasive properties
Microfiber™ - 80% polyester, 20% polyamide is highly absorbent and effective wet or dry

Ordering information: For other PolyMitt materials, contact your Avantor sales representative.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty