Would you like to log in again?
HiTrap™ TALON® crude Columns
Supplier: Cytiva
HiTrap™ Talon®
28-9537-67 28953767 28953766 29-0485-65 28-9537-66 29-0485-65
71006-000EA 1034.91 USD InStock
71006-000 CA11033-006 71005-998 CA71005-998 CA71006-000 89501-366
HiTrap™ TALON® crude Columns
Chromatography Columns
HiTrap TALON® crude 1 and 5 ml columns allow for rapid separations and easy scale-up with a minimum of sample preparation and equipment.
  • Columns prepacked with cobalt-based chromatography media designed for purification of histidine-tagged recombinant proteins by immobilized metal affinity chromatography (IMAC)
  • Suitable for IMAC purification of histidine-tagged proteins when Ni²⁺ is not the optimal choice of metal ion
  • Compatible with commonly used IMAC reagents and appropriate for purifying proteins under native or denaturing conditions
  • Prepacked HiTrap TALON® crude columns save time by allowing the application of unclarified samples directly, minimizing labor and degradation of proteins
  • HiTrap TALON® crude columns allow repeated runs without stripping, cleaning, or Co²⁺ recharging
TALON® Superflow™ is a cobalt-based IMAC medium offering different selectivity compared to nickel-charged media. TALON® Superflow™ and prepacked HiTrap TALON® crude columns allow protein purification under native or denaturing conditions and can be used with prokaryotic and eukaryotic expression systems. HiTrap TALON® crude columns allow for rapid separations and easy scale-up. Samples can be applied directly to the column after thorough cell disruption - without precentrifugation and filtration. The filters in the top and bottom of the column make it possible to directly load unclarified lysates on the column without causing back pressure problems or leakage of the TALON® Superflow™ beads. Prior to loading samples on HiTrap TALON® crude columns, cell lysate should be prepared using methods such as sonication, homogenization, or freeze/thaw.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty