Would you like to log in again?
GSTrap 4B Columns
Supplier: Cytiva
GSTrap™
28-4017-48 28-4017-47 28-4017-48 29-0486-09 28-4017-47 28-4017-45 29-0486-09 28-4017-45
95055-968PK 1957.01 USD InStock
CA95056-278L CA11033-014 CA95055-968L 95055-968 95056-050 CA95056-050L 89501-374 95056-278
GSTrap 4B Columns
Chromatography Columns
GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins.
  • High binding capacity
  • Mild elution conditions preserving protein antigenicity and function
  • Easy one-step purifications of Glutathione S-Transferase (GST) tagged proteins resulting in high purity
  • GSTrap 4B are designed to be used with a syringe, pump, or chromatography system, such as ÄKTA design
GST-tagged proteins can be purified directly from pretreated bacterial lysates using GSTrap 4B. GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids.
GST-tagged proteins can be detected using colorimetric or immunological methods. The resin, Glutathione Sepharose 4B, is also available as lab packages and is a good choice for scale-up. The columns can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÄKTA design or FPLC System.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty