Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HisTrap Columns, HP

Supplier: Cytiva
Histrap™
17-5247-01 17-5248-01 17-5248-02 17-5248-01 17-5248-02 17-5247-01 29-0510-21 29-0510-21
95056-206PK 945.54 USD InStock
CA95056-204L CA11033-028 CA95055-912L CA95056-206L 95056-206 95055-912 95056-204 89501-388
HisTrap Columns, HP
Chromatography Columns
HisTrap HP prepacked columns provide a fast, convenient and reproducible format for preparative purification of histidine-tagged (His-tag) recombinant proteins.

  • High resolution: Due to small bead size (average bead size is 34 µm)
  • High binding capacity: At least 40 mg His tag protein per mL resin, for high yields
  • Negligible nickel leakage: Helps retain activity and minimize protein precipitation
  • Broad reagent compatibility: Suitable for use with a wide range of reducing agents, detergents, denaturants, and other additives
  • Convenient HiTrap format: 1 ml and 5 ml columns compatible with syringe, pump, or chromatography systems such as ÄKTA or other FPLC systems

His tag protein purification by IMAC is both popular and highly efficient. HisTrap HP columns are packed with Ni Sepharose High Performance (HP) affinity resin. This resin consists of highly cross-linked agarose beads to which a chelating group has been coupled. The chelating group is precharged with nickel, which selectively retains proteins with exposed histidine groups.

HisTrap HP IMAC resin has a binding capacity that exceeds 40 mg/ml, resulting in high protein recovery. The small bead size (34 µm) contributes to a purer purification compared to nickel resins of larger bead size.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty